Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ FlashGel™ FLX Cassettes

Catalog No. BMA317258 Shop All Lonza, Inc Products
Change view
Click to view available options
Gel Percentage:
1.2%
2.2%
Wells:
12 + 1-well
2 x 16 + 1-well
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Percentage Wells
BMA317258 1.2% 12 + 1-well
BMA317260 2.2% 12 + 1-well
BMA317281 2.2% 2 x 16 + 1-well
BMA317259 1.2% 2 x 16 + 1-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BMA317258 Supplier Lonza™ Supplier No. 317258
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FlashGel™ FLX Cassettes are precast gels for use with the FlashGel™ System and support separation of DNA and RNA within 5 minutes.

FlashGel™ FLX Cassettes are precast gels for use with the FlashGel™ System. The FlashGel™ FLX Cassette supports the separation of either DNA or RNA, unlike standard FlashGel™ Cassettes. Band visualization is enabled through DNA- or RNA-specific stains included in the corresponding FlashGel™ FLX Prestain Loading Buffers. To separate DNA with FlashGel™ FLX Cassettes, use the FlashGel™ FLX DNA Prestain Loading Buffer (Cat. No. 316992) and to separate RNA, use the FlashGel™ FLX RNA Prestain Loading Buffer (Cat. No. 316992). Additionally, FlashGel™ FLX Cassettes offer an extended shelf life compared to standard FlashGel™ Cassettes, enhancing convenience and flexibility in routine workflows. FlashGel™ FLX Cassettes are fully compatible with the existing FlashGel™ System. They provide additional flexibility and convenience to the fastest system for separating DNA and RNA and monitoring band migration in real time. This highly sensitive system allows a 5 to 20X reduction in sample size - saving both time and money - completing separations within 5 minutes. Main benefits are:

  • Extended shelf life for reliable long-term use
  • Exceptional sensitivity and sharp resolution
  • Separation in as little as 5 minutes
  • Separate either DNA or RNA with same cassette
  • Real-time separation and instant documentation
  • Quick checks for enzymatic reactions (e.g., PCR amplification, restriction digestion, or T7 endonuclease cleavage)
  • Capture gel images directly at the bench - no need for UV transilluminators

Note that FlashGel™ FLX Cassettes are not recommended for recovery applications. Optimal separation range:

  • DNA: 100 bp - 10 kb separation range (separation of fragments >4kb will be improved by running longer at a lower voltage)
  • RNA: 0.5 - 9 kb separation range
TRUSTED_SUSTAINABILITY

Specifications

Compatible Ladders For DNA samples: FlashGel DNA Marker 100 bp to 4 kb For RNA samples: FlashGel RNA Marker 0.5 to 9 kb
Content And Storage Contains 9 gels Store gels at 18 to 26°C. Gels have a minimum shelf life of 6 months from the date of shipping.
Color Orange
Format Precast Gel
Gel Percentage 1.2%
Gel Size Mini
Gel Type Agarose
Label or Dye None
Sample Loading Volume 5 μL
Separation of DNA, RNA
Separation Range 100 to 10000 bp
Separation Type Electrophoresis
Wells 12 + 1-well
Gel Compatibility FlashGel System
Quantity 9 Gels/Pk.
Shipping Condition Room Temperature
Well Design Single-tier
Product Type Gel Cassette
For Use With (Equipment) FlashGel System
Show More Show Less

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.