Learn More
Lonza Walkersville FlashGel Visualization Glasses
Catalog No. 502120223
Encompass
FlashGel Visualization Glasses enable viewing of DNA bands to be recovered using the FlashGel Recovery System. The FlashGel Visualization Glasses enable direct viewing of DNA as it is being recovered. Using the FlashGel System users simply load samples, watch bands migrate and get data in as little as 2 minutes. Direct DNA recovery using The FlashGel Recovery System eliminates agarose gel preparation, band excision and purification. The system delivers highly efficient recovery, free from inhibitors and UV-induced damage, in a simple 5-10 minute protocol.Benefits:Directly visualize bands for recoverySee bands in as little as 2 minutesRecover samples directly without UV light band excision or purificationReal-time separation and documentationWatch band migration as it happensPhotograph gels at the bench without DNA damaging UV lightOutstanding sensitivity and resolution Applications:DNA ElectrophoresisRNA ElectrophoresisContent:FlashGel Visualization Glasses
Catalog No. 50-212-0223 Supplier Lonza Walkersville Supplier No. 57061
FlashGel Visualization Glasses enable viewing of DNA bands to be recovered using the FlashGel Recovery System. The FlashGel Visualization Glasses enable direct viewing of DNA as it is being recovered. Using the FlashGel System users simply load samples, watch bands migrate and get data in as little as 2 minutes. Direct DNA recovery using The FlashGel Recovery System eliminates agarose gel preparation, band excision and purification. The system delivers highly efficient recovery, free from inhibitors and UV-induced damage, in a simple 5-10 minute protocol.Benefits:Directly visualize bands for recoverySee bands in as little as 2 minutesRecover samples directly without UV light band excision or purificationReal-time separation and documentationWatch band migration as it happensPhotograph gels at the bench without DNA damaging UV lightOutstanding sensitivity and resolution Applications:DNA ElectrophoresisRNA ElectrophoresisContent:FlashGel Visualization Glasses