Learn More
Grainger Edwards Signaling Beacon, Flashing Incandescent, Red, 120 V AC, 1 ea
Catalog No. 502791713
Encompass_Preferred
The 51 Series flashing incandescent beacon with an integrated horn is a combination visual and audible signaling device engineered for both indoor and outdoor applications. Featuring a durable, shatter-resistant polycarbonate double fresnel lens, this beacon is designed to maximize light distribution and visibility. The cast base serves as a functional junction box, facilitating versatile installation in diverse environments.
  • Features an integrated horn synchronized with the flashing incandescent lamp
  • Operating temperature range of -35°C to 66°C
  • Includes a shatter-resistant double fresnel lens
  • Suitable for indoor and outdoor use
  • UL and cUL listed
  • Supports panel, conduit, or box mounting
  • Designed with an operating voltage of 120 V AC
Catalog No. 50-279-1713 Supplier Grainger Supplier No. 16G820
The 51 Series flashing incandescent beacon with an integrated horn is a combination visual and audible signaling device engineered for both indoor and outdoor applications. Featuring a durable, shatter-resistant polycarbonate double fresnel lens, this beacon is designed to maximize light distribution and visibility. The cast base serves as a functional junction box, facilitating versatile installation in diverse environments.
  • Features an integrated horn synchronized with the flashing incandescent lamp
  • Operating temperature range of -35°C to 66°C
  • Includes a shatter-resistant double fresnel lens
  • Suitable for indoor and outdoor use
  • UL and cUL listed
  • Supports panel, conduit, or box mounting
  • Designed with an operating voltage of 120 V AC