Learn More
Deltatrak Inc FlashLink Mini PDF IT Logger, 2/8ºC, 40 units per case
SDP
Catalog No. NC1730682
Encompass_Preferred
Preset with 5 minute sample intervals, two different models are offered. Model 31011 has 2C/8C low/high alarms for refrigerated conditions, model 31012 has 15C/25C alarms for CRT products. This plug-and-play logger has the added benefit of Shadow Logtechnology, guaranteeing that receivers always get trip data, even if a shipper forgets to start the unit. This compact, single-use temperature recorder has a built-in USB connector and on-board software, making it ideal for export shipments. No added software needed. Measurement Range -40F to 188.6F (-40C TO 87C) Accuracy 1.0F from -4F to 14F (0.5C from -20C to -10C) 0.6F from 14F to 50F (0.4C from -10C to 5C) 1.0F from 50F to 77F (0.5C from 5C to 25C) 1.8F if < -4F or > 77F (1.0C if < -20C or > 25C) Factory Programmable Alarms High & Low levels Battery Coin Cell Battery Certification CE Mark, Calibration traceable to NIST, RoHS Compliant Display Green and Red LED IP54
Catalog No. NC1730682 Supplier Deltatrak Inc Supplier No. 31011
Preset with 5 minute sample intervals, two different models are offered. Model 31011 has 2C/8C low/high alarms for refrigerated conditions, model 31012 has 15C/25C alarms for CRT products. This plug-and-play logger has the added benefit of Shadow Logtechnology, guaranteeing that receivers always get trip data, even if a shipper forgets to start the unit. This compact, single-use temperature recorder has a built-in USB connector and on-board software, making it ideal for export shipments. No added software needed. Measurement Range -40F to 188.6F (-40C TO 87C) Accuracy 1.0F from -4F to 14F (0.5C from -20C to -10C) 0.6F from 14F to 50F (0.4C from -10C to 5C) 1.0F from 50F to 77F (0.5C from 5C to 25C) 1.8F if < -4F or > 77F (1.0C if < -20C or > 25C) Factory Programmable Alarms High & Low levels Battery Coin Cell Battery Certification CE Mark, Calibration traceable to NIST, RoHS Compliant Display Green and Red LED IP54