Learn More
Deltatrak Inc FlashLink Mini PDF IT Logger 85 Days, 40 units per case
SDP
Catalog No. NC1730681
Encompass_Preferred
Our next generation plug-and-play logger has the added benefit of Shadow Logtechnology, guaranteeing that receivers always get trip data, even if a shipper forgets to start the unit. This compact, single-use temperature recorder has a built-in USB connector and on-board software, making it ideal for export shipments. No added software needed. Measurement Range -40F to 188.6F (-40C TO 87C) Accuracy 1.0F from -4F to 14F (0.5C from -20C to -10C) 0.6F from 14F to 50F (0.4C from -10C to 5C) 1.0F from 50F to 77F (0.5C from 5C to 25C) 1.8F if < -4F or > 77F (1.0C if < -20C or > 25C) Data Download Time 30 seconds typical Factory Programmable Alarms High & Low levels over entire range Delay Start 30 minutes Battery Coin Cell Battery Dimensions 1.858in x 3.071in x 0.402in (47.2mm x 78mm x 10.2mm) Weight 0.854 oz. (24.2 grams) Certification CE Mark, Calibration traceable to NIST, RoHS Compliant Display Green and Red LED IP54 with silicon cap
Catalog No. NC1730681 Supplier Deltatrak Inc Supplier No. 31010
Our next generation plug-and-play logger has the added benefit of Shadow Logtechnology, guaranteeing that receivers always get trip data, even if a shipper forgets to start the unit. This compact, single-use temperature recorder has a built-in USB connector and on-board software, making it ideal for export shipments. No added software needed. Measurement Range -40F to 188.6F (-40C TO 87C) Accuracy 1.0F from -4F to 14F (0.5C from -20C to -10C) 0.6F from 14F to 50F (0.4C from -10C to 5C) 1.0F from 50F to 77F (0.5C from 5C to 25C) 1.8F if < -4F or > 77F (1.0C if < -20C or > 25C) Data Download Time 30 seconds typical Factory Programmable Alarms High & Low levels over entire range Delay Start 30 minutes Battery Coin Cell Battery Dimensions 1.858in x 3.071in x 0.402in (47.2mm x 78mm x 10.2mm) Weight 0.854 oz. (24.2 grams) Certification CE Mark, Calibration traceable to NIST, RoHS Compliant Display Green and Red LED IP54 with silicon cap