Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI FlashPure EcoFlex C18 Functionalized Silica Bundle
SDP

Catalog No. 14000005K Shop All BUCHI Products
Change view
Click to view available options
Type:
50 μm Spherical Particles
51 μm Spherical Particles
Weight (Metric):
20 to 40 g
4 to 12 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Weight (Metric)
14-000-005K 50 μm Spherical Particles 20 to 40 g
14-000-004K 51 μm Spherical Particles 4 to 12 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-000-005K Supplier BUCHI Supplier No. 14000005K

missing translation for 'validMainframeMsg'

BUCHI FlashPure EcoFlex C18 Functionalized Silica Bundles are designed to optimized backpressure, price and performance

  • Ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Most economical solution without sacrificing quality
  • Large selection of cartridge sizes from 4g-3000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.

Specifications

Material Polyethylene
Weight (Metric) 20 to 40 g
Type 50 μm Spherical Particles
For Use With (Application) Non-Polar Compounds, Ideal for Peptides
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.