Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Silica
SDP

Catalog No. 05410011 Shop All BUCHI Products
Change view
Click to view available options
Includes:
1 Pc.
10 Pc.
12 Pc.
18 Pc.
2 Pc.
20 Pc.
3 Pc.
5 Pc.
6 Pc.
Bed Weight (Metric):
4 g
12 g
25 g
40 g
80 g
120 g
220 g
330 g
800 g
1600 g
3000 g
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Bed Weight (Metric)
05-410-011 3 Pc. 800 g
05-410-008 10 Pc. 120 g
05-410-004 18 Pc. 12 g
05-410-012 2 Pc. 1600 g
05-410-009 6 Pc. 220 g
05-410-005 12 Pc. 25 g
05-410-013 1 Pc. 3000 g
05-410-010 5 Pc. 330 g
05-410-006 12 Pc. 40 g
05-410-003 20 Pc. 4 g
05-410-007 10 Pc. 80 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 05-410-011 Supplier BUCHI Supplier No. 140000029
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.

  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters

Specifications

Material Polypropylene
Type Silica
Includes 3 Pc.
Resolution 40 to 63 μm
Bed Weight (Metric) 800 g
Column Type Silica
For Use With (Application) Basic Applications, Polar Compounds
For Use With (Equipment) Any Flash System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.