Learn More
BUCHI FlashPure Chromatography Solid Loader
SDP
Catalog No. 05410123 Shop All BUCHI Products
Change view
Click to view available options
Size (Metric) Tube Rack:
12 g
15 g
20 g
40 g
80 g
120 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Tube Rack
05-410-123 80 g
05-405-537 15 g
05-402-906 20 g
05-402-907 40 g
05-410-124 120 g
05-402-905 12 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 05-410-123 Supplier BUCHI Supplier No. 140000157
Only null left
Add to Cart
Add to Cart
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Material Polypropylene
Style Frits
Type Empty
Includes Frits
Size (Metric) Tube Rack 80 g
Column Type Empty
For Use With (Application) Solid Loading of Insoluble Samples
For Use With (Equipment) Reveleris Solid Loading