Learn More
BUCHI FlashPure Empty Solid Loader: Parts
SDP
Catalog No. 05410137 Shop All BUCHI Products
Change view
Click to view available options
FlashPure Empty Solid Loader Frits; Size: 40 g; Quantity: 10 pcs
Catalog No.
05-410-137 FlashPure Empty Solid Loader Frits; Size: 40 g; Quantity: 10 pcs
1 options
Catalog No. 05-410-137 Supplier BUCHI Supplier No. 148634560
Add to Cart
Add to Cart
Parts for FlashPure Empty Solid Loaders. FlashPure Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Material Polyethylene
Type Frit
Includes 10 Pc.
Size (Metric) Tube Rack 40 g
Column Type Empty
For Use With (Application) Solid Loading of Insoluble Samples
For Use With (Equipment) Reveleris Solid Loading