Learn More
BUCHI FlashPure Empty Solid Loader: Parts
SDP
Catalog No. 05410133 Shop All BUCHI Products
Change view
Click to view available options
FlashPure Empty Solid Loader Insertion Rod; Size: 12-20 g; Quantity: 1 pc
Catalog No.
05-410-133 FlashPure Empty Solid Loader Insertion Rod; Size: 12-20 g; Quantity: 1 pc
1 options
Catalog No. 05-410-133 Supplier BUCHI Supplier No. 142129062
Parts for FlashPure Empty Solid Loaders. FlashPure Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Material Polypropylene
Type Insertion Rod
Includes 1 Pc.
Size (Metric) Tube Rack 12 to 20 g
For Use With (Application) Solid Loading of Insoluble Samples
For Use With (Equipment) Reveleris Solid Loading