Learn More
BUCHI FlashPure Empty Solid Loader Bundle
SDP
Catalog No. 1486343K Shop All BUCHI Products
Change view
Click to view available options
FlashPure Empty Solid Loader Bundle
Catalog No.
14-863-43K FlashPure Empty Solid Loader Bundle
1 options
Catalog No. 14-863-43K Supplier BUCHI Supplier No. 1486343K
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
BUCHI FlashPure Empty Solid Loader Bundle is ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Material Polyethylene
Weight (Metric) 12 to 40 g
Type Empty
For Use With (Application) Solid Loading of Insoluble Samples