Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI FlashPure Select Flash Chromatography Cartridges: C18
SDP

Catalog No. 05405552 Shop All BUCHI Products
Change view
Click to view available options
Bed Weight (Metric):
800 g
1500 g
3000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight (Metric)
05-405-552 3000 g
05-405-550 800 g
05-405-551 1500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-405-552 Supplier BUCHI Supplier No. 11070492
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for ultimate and unmatched performance.

  • Spherical particles
  • 13-17 μm silica, 20-35 μm C18
  • Functionalized silica cartridges can be reused multiple times
  • Large selection of cartridge sizes from 4 g - 3000 g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.

Specifications

Material Polypropylene
Type Flash Column
Resolution 20 to 35 μm
Bed Weight (Metric) 3000 g
Column Type C18
For Use With (Application) Non-Polar Compounds, Ideal for Peptides
For Use With (Equipment) Any Flash System
Quantity 1 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.