Learn More
Ace Glass, Inc. Flask, 10L, rotary evaporator, receiver, for 20L Heidolph units, 40/25 center, GL18 side neck with cap, 0-10mm GL18bottom outlet with 3/8in hose connection. Replaces part number 036303040.
SDP
Catalog No. 50978852
Encompass
Flask, 10L, rotary evaporator, receiver, for 20L Heidolph units, 40/25 center, GL18 side neck with cap, 0-10mm GL18bottom outlet with 3/8in hose connection. Replaces part number 036303040.
Catalog No. 50-978-852 Supplier Ace Glass, Inc. Supplier No. 670144
Flask, 10L, rotary evaporator, receiver, for 20L Heidolph units, 40/25 center, GL18 side neck with cap, 0-10mm GL18bottom outlet with 3/8in hose connection. Replaces part number 036303040.