Learn More
Triforest Enterprises 125ML sterile baffled bottom erlenmeyer shaker flask
Catalog No. NC2005099
Encompass_Preferred
TriForest Shaker Flasks are manufactured with virgin, optically clear, non leaching polycarbonate resin that retains clarity after gamma radiation. The flasks are certified pyrogen free, DNase/RNase free, ideal for cell cultures, microbial cultures, plants, media preparation, storage, and all related applications. They fit all standard shaking incubator clamps and can be placed on any platform.
Catalog No. NC2005099 Supplier Triforest Enterprises Supplier No. FBC0125S
TriForest Shaker Flasks are manufactured with virgin, optically clear, non leaching polycarbonate resin that retains clarity after gamma radiation. The flasks are certified pyrogen free, DNase/RNase free, ideal for cell cultures, microbial cultures, plants, media preparation, storage, and all related applications. They fit all standard shaking incubator clamps and can be placed on any platform.