Learn More
Ace Glass, Inc. Flask, reaction, jacketed, 12L, 6in grooved flange, 28/15 O-ring ball joints on jacket inlet/outlets
SDP
Catalog No. 50973219
Encompass
12L jacketed reaction flask with a 6 grooved flange, 28/15 o-ring ball joints on the jacket inlet/outlets, and reference graduations. Flask should be used with a stainless steel quick release clamp. Supplied with one CAPFE o-ring (PTFE encapsulated silicone).
Catalog No. 50-973-219 Supplier Ace Glass, Inc. Supplier No. 653512
12L jacketed reaction flask with a 6 grooved flange, 28/15 o-ring ball joints on the jacket inlet/outlets, and reference graduations. Flask should be used with a stainless steel quick release clamp. Supplied with one CAPFE o-ring (PTFE encapsulated silicone).