Learn More
CHEMSCIENCE Volumetric Flask, 1500mL, Serialized and Certified, Heavy Duty, Wide Mouth, PE Stopper# 27, Class A,PK/2
Catalog No. NC2681271
Encompass_Preferred
Wide mouth design facilitates mixing, pipette access, sampling and filling. Unique flat bottom interior permits the use of a standard stir bar. The graduation line on the neck is sharp with permanent large colored encryption for easy-read. Flasks are calibrated 'To Contain' at Class A tolerances, per ASTM E288 specifications. Supplied with closed bottom Polyethylene stopper to fit in the ground neck.
Catalog No. NC2681271 Supplier CHEMSCIENCE Supplier No. 129.503.10A
Wide mouth design facilitates mixing, pipette access, sampling and filling. Unique flat bottom interior permits the use of a standard stir bar. The graduation line on the neck is sharp with permanent large colored encryption for easy-read. Flasks are calibrated 'To Contain' at Class A tolerances, per ASTM E288 specifications. Supplied with closed bottom Polyethylene stopper to fit in the ground neck.