Learn More
Chemglass Life Sciences 250mL Apollo Flask, 24/40 Outer Joint
SDP
Catalog No. CG1512A06
Encompass_Preferred
250mL Apollo Flask, 24/40 Outer Joint. HEAVY WALL single neck flask with a standard taper outer joint. Low profile design allows the flask to sit upright on the bench and also permits easy access of products with a spatula or policeman. The flask contour is excellent for controlled evaporation of foaming solvents during rotary evaporation.
Catalog No. CG1512A06 Supplier Chemglass Life Sciences Supplier No. CG1512A06
250mL Apollo Flask, 24/40 Outer Joint. HEAVY WALL single neck flask with a standard taper outer joint. Low profile design allows the flask to sit upright on the bench and also permits easy access of products with a spatula or policeman. The flask contour is excellent for controlled evaporation of foaming solvents during rotary evaporation.