Learn More
Ace Glass, Inc. Flask, 1L, 4in O-ring grooved flange, 0-10mm ZDS bottom valve
SDP
Catalog No. 50973138
Encompass_Preferred
1L cylindrical reaction flask with grooved flange opening. Flask has Zero Dead Space bottom valve. Valve features no material hang-up in open position. When closed, the valve sits flush with bottom of the flask. Includes one CAPFE o-ring.
Catalog No. 50-973-138 Supplier Ace Glass, Inc. Supplier No. 651810
1L cylindrical reaction flask with grooved flange opening. Flask has Zero Dead Space bottom valve. Valve features no material hang-up in open position. When closed, the valve sits flush with bottom of the flask. Includes one CAPFE o-ring.