Learn More
Ace Glass, Inc. Flask, reaction, 1L, jacketed, 137mm O-ring groove flat flange, 140mm bottom OD x 165mm H, #11 Ace-Thred joints on jacket inlet/outlet for use with 3/8in ID tubing
SDP
Catalog No. 50978788
Encompass_Preferred
1L jacketed reaction flask with round bottom, and #11 Ace-Thred joints on jacket inlet/outlet for use with 3/8 I.D. tubing. 137mm o-ring groove flat flange is used to seal a head, instead of a flat gasket. Jacketed for temperature control of ingredients. Supplied with CAPFE o-rings. Dimensions: 140mm bottom O.D. x 165mm H.
Catalog No. 50-978-788 Supplier Ace Glass, Inc. Supplier No. 6519301
1L jacketed reaction flask with round bottom, and #11 Ace-Thred joints on jacket inlet/outlet for use with 3/8 I.D. tubing. 137mm o-ring groove flat flange is used to seal a head, instead of a flat gasket. Jacketed for temperature control of ingredients. Supplied with CAPFE o-rings. Dimensions: 140mm bottom O.D. x 165mm H.