Learn More
Ace Glass, Inc. Flask, reaction, 200L, 200mm(8in) ground Duran; O-ring groove flange, flush-seal bottom valve
SDP
Catalog No. 50973162
Encompass
200L cylindrical reaction flask with Duran flat-ground 200mm(8) o-ring groove flange, and a flush-seal bottom valve. For use with a stainless steel quick release clamp. The flask is supplied with a CAPFE o-ring.
Catalog No. 50-973-162 Supplier Ace Glass, Inc. Supplier No. 652286
Add to Cart
Add to Cart
Request a Quote
200L cylindrical reaction flask with Duran flat-ground 200mm(8) o-ring groove flange, and a flush-seal bottom valve. For use with a stainless steel quick release clamp. The flask is supplied with a CAPFE o-ring.