Learn More
Ace Glass, Inc. Flask, round bottom, 250mL, (2)24/40 necks, 0-3mm inlet valve sidearm, No-Air
SDP
Catalog No. 50975433
Encompass_Preferred
250mL No-Air round bottom flask with (2) 24/40 standard taper necks, and a 0-3mm inlet valve sidearm with an 8mm end for use with a rubber septa. Excellent flask for reactions, and No-Air or vacuum applications.
Catalog No. 50-975-433 Supplier Ace Glass, Inc. Supplier No. 7799112
250mL No-Air round bottom flask with (2) 24/40 standard taper necks, and a 0-3mm inlet valve sidearm with an 8mm end for use with a rubber septa. Excellent flask for reactions, and No-Air or vacuum applications.