Learn More
Ace Glass, Inc. Flask, round bottom, 250mL, 24/40 neck, 2mm glass stopcock sidearm, includes septa, No-Air
SDP
Catalog No. 50973997
Encompass_Preferred
250mL round bottom flask with a single neck with a 24/40 standard taper joint, and an 8mm sidearm with a 2mm glass stopcock. Ideal for Schlenk lines and either NO-AIR or vacuum applications. Includes (1) rubber septa for the sidearm.
Catalog No. 50-973-997 Supplier Ace Glass, Inc. Supplier No. 700325
250mL round bottom flask with a single neck with a 24/40 standard taper joint, and an 8mm sidearm with a 2mm glass stopcock. Ideal for Schlenk lines and either NO-AIR or vacuum applications. Includes (1) rubber septa for the sidearm.