Learn More
Ace Glass, Inc. Flask, 250mL, filter, #11 Ace-Thred hose connection
SDP
Supplier:  Ace Glass, Inc. 698308
Encompass_Preferred
250mL heavy wall filter flask with a tooled neck for uniform stopper fit. Side hose connection is a #11 Ace-Thred that accepts a PTFE tubing connector with a silicone o-ring and nylon bushing to make an easier, safer, more convenient vacuum hose connection. Flask only; connector not included.
Catalog No. 50-973-973