Learn More
Thomson Instrument Company Ultra Yield 250mL Flask, Sterile
SDP
Catalog No. 50996059
Encompass_Preferred
Ultra Yield Flasks are sterile, single-use shake flasks engineered for max oxygen transfer and high-density growth. Their high-aeration geometry, used on standard orbital shakers, drives faster cultures and cleaner downstreams. Pair with AirOtop Enhanced Seals or Vented Screw Caps; with Plasmid+ Media, customers have reported plasmid yields up to 610%.Key features
•High-aeration geometry/baffled base to boost OTR and mixing
•Sterile, ready-to-use single-use format for low contamination risk
•Sizes from 125 mL to 2.5 L for bench-to-pilot scalability
•Engineered for high-rpm orbital shaking (stable, leak-tight operation)
•Closure flexibility: AirOtop seals, Vented Screw Caps, or Bidirectional Transfer Caps
•Clear graduations & label area for easy sampling and tracking
•Made in the USA with reliable, fast shipment
Catalog No. 50-996-059 Supplier Thomson Instrument Company Supplier No. 931144
Ultra Yield Flasks are sterile, single-use shake flasks engineered for max oxygen transfer and high-density growth. Their high-aeration geometry, used on standard orbital shakers, drives faster cultures and cleaner downstreams. Pair with AirOtop Enhanced Seals or Vented Screw Caps; with Plasmid+ Media, customers have reported plasmid yields up to 610%.Key features
•High-aeration geometry/baffled base to boost OTR and mixing
•Sterile, ready-to-use single-use format for low contamination risk
•Sizes from 125 mL to 2.5 L for bench-to-pilot scalability
•Engineered for high-rpm orbital shaking (stable, leak-tight operation)
•Closure flexibility: AirOtop seals, Vented Screw Caps, or Bidirectional Transfer Caps
•Clear graduations & label area for easy sampling and tracking
•Made in the USA with reliable, fast shipment