Learn More
Chemglass Life Sciences Flask, Fernbach, 2800mL, GL-45 Thread, 3 Side Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230
SDP
Catalog No. 501532256
Encompass_Preferred
Flask, Fernbach, 2800mL, GL-45 Thread, 3 Side Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Flask has 3 baffles on lower outside edge to help oxygen transfer to the culture medium.
Catalog No. 50-153-2256 Supplier Chemglass Life Sciences Supplier No. CLS202231
Flask, Fernbach, 2800mL, GL-45 Thread, 3 Side Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Flask has 3 baffles on lower outside edge to help oxygen transfer to the culture medium.