Learn More
Ace Glass, Inc. Flask, reaction, 2L, 100mm(4in) ground Duran; flange -ring groove
SDP
Catalog No. 50973149
Encompass_Preferred
2L reaction cylindrical reaction flask with 100mm (4 inch) Schott ground flange with o-ring groove, for use with stainless steel quick release clamp (part number 509557344). Flask is supplied with one CAPFE (PTFE encapsulated silicone) o-ring.
Catalog No. 50-973-149 Supplier Ace Glass, Inc. Supplier No. 652112
2L reaction cylindrical reaction flask with 100mm (4 inch) Schott ground flange with o-ring groove, for use with stainless steel quick release clamp (part number 509557344). Flask is supplied with one CAPFE (PTFE encapsulated silicone) o-ring.