Learn More
Ace Glass, Inc. Flask, reaction, 2L, jacketed, 168mm O-ring groove flat flange, 170mm bottom OD x 185mm H, #11 Ace-Thred joints on jacket inlet/outlet for use with 3/8in ID tubing
SDP
Catalog No. 50973146
Encompass_Preferred
2L jacketed reaction flask with round bottom, and #11 Ace-Thred joints on jacket inlet/outlet for use with 3/8 I.D. tubing. 168mm o-ring groove flat flange is used to seal a head, instead of a flat gasket. Jacketed for temperature control of ingredients. Supplied with CAPFE o-rings. Dimensions: 170mm bottom O.D. x 185mm H.
Catalog No. 50-973-146 Supplier Ace Glass, Inc. Supplier No. 6519302
2L jacketed reaction flask with round bottom, and #11 Ace-Thred joints on jacket inlet/outlet for use with 3/8 I.D. tubing. 168mm o-ring groove flat flange is used to seal a head, instead of a flat gasket. Jacketed for temperature control of ingredients. Supplied with CAPFE o-rings. Dimensions: 170mm bottom O.D. x 185mm H.