Learn More
Cole-Parmer Labconco Fast-Freeze™ 7540600 Clear Glass Drying Flask; 300 mL
Catalog No. NC2118060 Shop All Cole-Parmer Products
Encompass
Provides easy and efficient handling of freeze dry samplesBorosilicate glass withstands extreme temperatures and high vacuumCompatible with any brand freeze dryer with ¾” diameter valve portsFlat bottom and wide mouth design provides easy loading and cleaningThe Fast-Freeze™ drying flask is constructed of high strength borosilicate glass which can withstand a maximum temperature of 490º C. It easily attaches to the valve ports on Labconco drying chambers, manifolds, and freeze dryer systems with the use of an adapter (sold separately).
This item is not returnable. View return policy
Catalog No. NC2118060 Supplier Cole-Parmer Supplier No. 0335304
Provides easy and efficient handling of freeze dry samplesBorosilicate glass withstands extreme temperatures and high vacuumCompatible with any brand freeze dryer with ¾” diameter valve portsFlat bottom and wide mouth design provides easy loading and cleaningThe Fast-Freeze™ drying flask is constructed of high strength borosilicate glass which can withstand a maximum temperature of 490º C. It easily attaches to the valve ports on Labconco drying chambers, manifolds, and freeze dryer systems with the use of an adapter (sold separately).