Learn More
Ace Glass, Inc. Flask, 30mL, 14/20, round bottom, micro Soxhlet
SDP
Catalog No. 50978879
Encompass_Preferred
30mL round bottom flask with 14/20 standard taper joint, for use with ACE micro Soxhlet extractor (50973419). Soxhlet extractor is designed for small chemical applications. Improved design permits greater condensing capacity.
Catalog No. 50-978-879 Supplier Ace Glass, Inc. Supplier No. 677606
30mL round bottom flask with 14/20 standard taper joint, for use with ACE micro Soxhlet extractor (50973419). Soxhlet extractor is designed for small chemical applications. Improved design permits greater condensing capacity.