Learn More
Ace Glass, Inc. Flask, 3L, spherical, jacketed, 100mm O-ring groove flange, 0-8mm bottom, #11 Ace-Thred inlet/outlets for use with 3/8inid tubing
SDP
Catalog No. 50973224
Encompass_Preferred
Flask, 3L, spherical, jacketed, 100mm O-ring groove flange, 0-8mm bottom, #11 Ace-Thred inlet/outlets for use with 3/8inid tubing
Catalog No. 50-973-224 Supplier Ace Glass, Inc. Supplier No. 6536306
Flask, 3L, spherical, jacketed, 100mm O-ring groove flange, 0-8mm bottom, #11 Ace-Thred inlet/outlets for use with 3/8inid tubing