Learn More
Ace Glass, Inc. Flask, jacketed, 3L, 100mm(4in) ground Duran; O-ring groove flange, 0-8mm easy-action bottom valve with 28/15o-ring ball joint, 28/15 O-ring ball joint jacket inlet/outlet
SDP
Catalog No. 50973173
Encompass_Preferred
3L jacketed reaction flask with a 100mm (4) o-ring groove flange, and a 0-8mm easy-action bottom valve with a 28/15 o-ring ball joint. The jacket inlet/outlets have tangentially sealed 28/15 o-ring ball joints.
Catalog No. 50-973-173 Supplier Ace Glass, Inc. Supplier No. 652417
3L jacketed reaction flask with a 100mm (4) o-ring groove flange, and a 0-8mm easy-action bottom valve with a 28/15 o-ring ball joint. The jacket inlet/outlets have tangentially sealed 28/15 o-ring ball joints.