Learn More
Ace Glass, Inc. Flask, jacketed, 3L, 100mm(4in) ground Duran; O-ring groove flange, 0-8mm easy-action bottom valve with 28/15o-ring ball joint, #11 Ace-Thred™; jacket inlet/outlet for 3/8in ID tubing
SDP
Catalog No. 50978798
Encompass_Preferred
3L jacketed reaction flask with a 100mm (4) o-ring groove flange, and a 0-8mm easy-action bottom valve with a 28/15 o-ring ball joint. The jacket inlet/outlets have #11 Ace-Thred connections for use with 3/8 I.D. tubing.
Catalog No. 50-978-798 Supplier Ace Glass, Inc. Supplier No. 6524317
3L jacketed reaction flask with a 100mm (4) o-ring groove flange, and a 0-8mm easy-action bottom valve with a 28/15 o-ring ball joint. The jacket inlet/outlets have #11 Ace-Thred connections for use with 3/8 I.D. tubing.