Learn More
Ace Glass, Inc. Flask, 4L, 6in O-ring grooved flange, 0-20mm ZDS bottom valve
SDP
Catalog No. 50973142
Encompass_Preferred
4, 6in cylindrical reaction flask with grooved flange opening. Flask has Zero Dead Space bottom valve. Valve features no material hang-up in open position. When closed, the valve sits flush with the bottom of the flask. Includes one CAPFE o-ring.
Catalog No. 50-973-142 Supplier Ace Glass, Inc. Supplier No. 651820
4, 6in cylindrical reaction flask with grooved flange opening. Flask has Zero Dead Space bottom valve. Valve features no material hang-up in open position. When closed, the valve sits flush with the bottom of the flask. Includes one CAPFE o-ring.