Learn More
Ace Glass, Inc. Flask, reaction, 4L, jacketed, 168mm O-ring groove flat flange, 140mm bottom OD x 335mm H, 28/15 O-ring ball joints on jacket inlet/outlet
SDP
Catalog No. 50973145
Encompass_Preferred
4L jacketed reaction flask with round bottom and 28/15 o-ring ball joints on jacket inlet/outlet. 168mm o-ring groove flat flange is used to seal a head, instead of a flat gasket. Jacketed for temperature control of ingredients. Supplied with CAPFE o-rings. Dimensions: 140mm bottom O.D. x 335mm H.
Catalog No. 50-973-145 Supplier Ace Glass, Inc. Supplier No. 651904
4L jacketed reaction flask with round bottom and 28/15 o-ring ball joints on jacket inlet/outlet. 168mm o-ring groove flat flange is used to seal a head, instead of a flat gasket. Jacketed for temperature control of ingredients. Supplied with CAPFE o-rings. Dimensions: 140mm bottom O.D. x 335mm H.