Learn More
Ace Glass, Inc. Flask, round bottom, 500mL, (2)24/40 necks, 2mm glass stopcock sidearm, No-Air
SDP
Catalog No. 50975431
Encompass_Preferred
500mL No-Air round bottom flask with (2) 24/40 standard taper necks, and a 2mm glass stopcock sidearm. Excellent flask for reactions, and No-Air or vacuum applications.
Catalog No. 50-975-431 Supplier Ace Glass, Inc. Supplier No. 779908
500mL No-Air round bottom flask with (2) 24/40 standard taper necks, and a 2mm glass stopcock sidearm. Excellent flask for reactions, and No-Air or vacuum applications.