Learn More
Ace Glass, Inc. FLASK,50L,200MM,FLUSH BOTTOM
SDP
Catalog No. 5000854709
Encompass
Product family consisting of large spherical flasks and the heads that complement them. Flasks that feature a bottom outlet have descriptions that state the outlet's type, otherwise the flask has no outlet.
Catalog No. 50-008-54709 Supplier Ace Glass, Inc. Supplier No. 653014
Product family consisting of large spherical flasks and the heads that complement them. Flasks that feature a bottom outlet have descriptions that state the outlet's type, otherwise the flask has no outlet.