Learn More
Ace Glass, Inc. Flask, round bottom, 50mL, (2)14/20 necks, 0-3mm inlet valve sidearm, No-Air
SDP
Catalog No. 50979465
Encompass_Preferred
50mL No-Air round bottom flask with (2) 14/20 standard taper necks, and a 0-3mm inlet valve sidearm with an 8mm end for use with a rubber septa. Excellent flask for reactions, and No-Air or vacuum applications.
Catalog No. 50-979-465 Supplier Ace Glass, Inc. Supplier No. 7799100
50mL No-Air round bottom flask with (2) 14/20 standard taper necks, and a 0-3mm inlet valve sidearm with an 8mm end for use with a rubber septa. Excellent flask for reactions, and No-Air or vacuum applications.