Learn More
Ace Glass, Inc. Flask, round bottom, 50mL, 0-4mm Hi-Vac valve top with a 15mmo-ring joint sidearm, No-Air
SDP
Catalog No. 50979473
Encompass_Preferred
Flask, round bottom, 50mL, 0-4mm Hi-Vac valve top with a 15mmo-ring joint sidearm, No-Air
Catalog No. 50-979-473 Supplier Ace Glass, Inc. Supplier No. 781602
Flask, round bottom, 50mL, 0-4mm Hi-Vac valve top with a 15mmo-ring joint sidearm, No-Air