Learn More
Ace Glass, Inc. Flask, reaction, jacketed, 5L, 4in grooved flange, 28/15 O-ring ball joints on jacket inlet/outlets
SDP
Catalog No. 50978812
Encompass_Preferred
5L jacketed reaction flask with a 4 grooved flange, and 28/15 o-ring ball joints on the jacket inlet/outlets. Flask should be used with a stainless steel quick release clamp. Supplied with one CAPFE o-ring (PTFE encapsulated silicone).
Catalog No. 50-978-812 Supplier Ace Glass, Inc. Supplier No. 653505
5L jacketed reaction flask with a 4 grooved flange, and 28/15 o-ring ball joints on the jacket inlet/outlets. Flask should be used with a stainless steel quick release clamp. Supplied with one CAPFE o-ring (PTFE encapsulated silicone).