Learn More
Ace Glass, Inc. Flask, reaction, jacketed, 5L, 4in grooved flange, #11 Ace-Thred connections on jacket inlet/outlets for use with 3/8in
SDP
Catalog No. 50978813
Encompass_Preferred
5L jacketed reaction flask with a 4 grooved flange, and #11 Ace-Thred connections on the jacket inlet/outlets for use with 3/8 I.D. tubing. Flask should be used with a stainless steel quick release clamp. Supplied with one CAPFE o-ring (PTFE encapsulated silicone).
Catalog No. 50-978-813 Supplier Ace Glass, Inc. Supplier No. 6535305
5L jacketed reaction flask with a 4 grooved flange, and #11 Ace-Thred connections on the jacket inlet/outlets for use with 3/8 I.D. tubing. Flask should be used with a stainless steel quick release clamp. Supplied with one CAPFE o-ring (PTFE encapsulated silicone).