Learn More
Ace Glass, Inc. Flask, 5L, spherical, jacketed, 150mm O-ring groove flange, 0-8mm bottom, #11 Ace-Thred inlet/outlets for use with 3/8inid tubing
SDP
Catalog No. 50973226
Encompass_Preferred
Flask, 5L, spherical, jacketed, 150mm O-ring groove flange, 0-8mm bottom, #11 Ace-Thred inlet/outlets for use with 3/8inid tubing
Catalog No. 50-973-226 Supplier Ace Glass, Inc. Supplier No. 6536311
Flask, 5L, spherical, jacketed, 150mm O-ring groove flange, 0-8mm bottom, #11 Ace-Thred inlet/outlets for use with 3/8inid tubing