Learn More
Ace Glass, Inc. Flask, 5L, spherical, jacketed, 150mm O-ring groove flange, 0-8mm bottom, 28/15 ball joint inlet/outlets
SDP
Catalog No. 50978814
Encompass_Preferred
Flask, 5L, spherical, jacketed, 150mm O-ring groove flange, 0-8mm bottom, 28/15 ball joint inlet/outlets
Catalog No. 50-978-814 Supplier Ace Glass, Inc. Supplier No. 653611
Flask, 5L, spherical, jacketed, 150mm O-ring groove flange, 0-8mm bottom, 28/15 ball joint inlet/outlets