Learn More
Ace Glass, Inc. Flask, reaction, 5L, indents, 6in grooved flange
SDP
Catalog No. 50973228
Encompass_Preferred
5L rugged spherical reaction flask with indents for greater agitation. Flask has a 6 Duran flat-ground flange with an O-ring groove, for use with a stainless steel quick release clamp. Flask is supplied with one CAPFE O-ring; CAPFE is PTFE encapsulated silicone.
Catalog No. 50-973-228 Supplier Ace Glass, Inc. Supplier No. 653717
5L rugged spherical reaction flask with indents for greater agitation. Flask has a 6 Duran flat-ground flange with an O-ring groove, for use with a stainless steel quick release clamp. Flask is supplied with one CAPFE O-ring; CAPFE is PTFE encapsulated silicone.