Learn More
Ace Glass, Inc. Flask, reaction, 6L, 200mm(8in) ground Duran; O-ring groove flange, 0-8mm easy-action bottom valve with 28/15o-ring ball joint connection
SDP
Catalog No. 50978792
Encompass_Preferred
6L cylindrical reaction flask with Duran flat-ground 200mm(8) o-ring groove flange, and a 0-8 Easy Action Plug bottom valve with 28/15 o-ring ball joint connection with CHEMRAZ o-rings for draining contents. For use with a stainless steel quick release clamp. The flask is supplied with a CAPFE o-ring.
Catalog No. 50-978-792 Supplier Ace Glass, Inc. Supplier No. 652217
6L cylindrical reaction flask with Duran flat-ground 200mm(8) o-ring groove flange, and a 0-8 Easy Action Plug bottom valve with 28/15 o-ring ball joint connection with CHEMRAZ o-rings for draining contents. For use with a stainless steel quick release clamp. The flask is supplied with a CAPFE o-ring.