Learn More
Ace Glass, Inc. Flask, jacketed, 6L, 200mm(8in) ground Duran; O-ring groove flange, 0-8mm easy-action bottom valve with 28/15o-ring ball joint, #15 Ace-Thred™; jacket inlet/outlet for 1/2in ID tubing
SDP
Catalog No. 50973176
Encompass
6L jacketed reaction flask with a 200mm (8) o-ring groove flange, and a 0-8mm easy-action bottom valve with a 28/15 o-ring ball joint. The jacket inlet/outlets have #15 Ace-Thred connections for use with 1/2 I.D. tubing.
Catalog No. 50-973-176 Supplier Ace Glass, Inc. Supplier No. 6524320
6L jacketed reaction flask with a 200mm (8) o-ring groove flange, and a 0-8mm easy-action bottom valve with a 28/15 o-ring ball joint. The jacket inlet/outlets have #15 Ace-Thred connections for use with 1/2 I.D. tubing.