Learn More
Ace Glass, Inc. Flask, reaction, 6L, 200mm(8in) ground Duran; flange -ring groove
SDP
Catalog No. 50973151
Encompass_Preferred
6L reaction cylindrical reaction flask with 200mm (8 inch) Schott ground flange with o-ring groove, for use with stainless steel quick release clamp (part number 50978785). Flask is supplied with one CAPFE (PTFE encapsulated silicone) o-ring.
Catalog No. 50-973-151 Supplier Ace Glass, Inc. Supplier No. 652116
6L reaction cylindrical reaction flask with 200mm (8 inch) Schott ground flange with o-ring groove, for use with stainless steel quick release clamp (part number 50978785). Flask is supplied with one CAPFE (PTFE encapsulated silicone) o-ring.