Learn More
Chemglass Life Sciences 250mL Apollo Flask, 14/20 Outer Joint
SDP
Catalog No. 501531134
Encompass_Preferred
250mL Apollo Flask, 14/20 Outer Joint. HEAVY WALL single neck flask with a standard taper outer joint. Low profile design allows the flask to sit upright on the bench and also permits easy access of products with a spatula or policeman. The flask contour is excellent for controlled evaporation of foaming solvents during rotary evaporation.
Catalog No. 50-153-1134 Supplier Chemglass Life Sciences Supplier No. CG1512A05
250mL Apollo Flask, 14/20 Outer Joint. HEAVY WALL single neck flask with a standard taper outer joint. Low profile design allows the flask to sit upright on the bench and also permits easy access of products with a spatula or policeman. The flask contour is excellent for controlled evaporation of foaming solvents during rotary evaporation.