Learn More
Chemglass Life Sciences 2000mL Glassblowers Erlenmeyer Blank
SDP
Catalog No. 501531805
Encompass_Preferred
2000mL Glassblowers Erlenmeyer Blank. Blanks for fabrication of Erlenmeyer shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. 50-153-1805 Supplier Chemglass Life Sciences Supplier No. CG62008
2000mL Glassblowers Erlenmeyer Blank. Blanks for fabrication of Erlenmeyer shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.