Learn More
Chemglass Life Sciences 125mL Glassblowers Erlenmeyer Blank
SDP
Catalog No. 50873568
Encompass_Preferred
125mL Glassblowers Erlenmeyer Blank. Blanks for fabrication of Erlenmeyer shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. 50-873-568 Supplier Chemglass Life Sciences Supplier No. CG62004
125mL Glassblowers Erlenmeyer Blank. Blanks for fabrication of Erlenmeyer shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.