Learn More
Chemglass Life Sciences Flask, Fernbach, 2800mL, Plain 59mm ID Top, WITHOUT Baffles, Approx OD x Height (mm): 210 x 230
SDP
Catalog No. 501532255
Encompass_Preferred
Flask, Fernbach, 2800mL, Plain 59mm ID Top, WITHOUT Baffles, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Available with a plain 59mm ID neck.
Catalog No. 50-153-2255 Supplier Chemglass Life Sciences Supplier No. CLS202010
Flask, Fernbach, 2800mL, Plain 59mm ID Top, WITHOUT Baffles, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Available with a plain 59mm ID neck.